Skip to Content
Merck
All Photos(1)

Documents

320366

Sigma-Aldrich

Isopropylamine

99%

Synonym(s):

2-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
605259
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.04 (vs air)

vapor pressure

9.2 psi ( 20 °C)

Assay

99%

autoignition temp.

755 °F

expl. lim.

10.4 %

refractive index

n20/D 1.374 (lit.)

bp

33-34 °C (lit.)

mp

−101 °C

density

0.688 g/mL at 20 °C (lit.)
0.694 g/mL at 25 °C

SMILES string

CC(C)N

InChI

1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3

InChI key

JJWLVOIRVHMVIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 471291-ALDRICH | Isopropylamine ≥99.5%

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-13.0 °F - closed cup

Flash Point(C)

<= -25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Donna L McGovern et al.
Journal of molecular graphics & modelling, 28(7), 612-625 (2010-01-20)
The highly potent and kappa-opioid (KOP) receptor-selective hallucinogen Salvinorin A and selected analogs have been analyzed using the 3D quantitative structure-affinity relationship technique Comparative Molecular Field Analysis (CoMFA) in an effort to derive a statistically significant and predictive model of
Chaw Jiang Lim et al.
Pest management science, 69(1), 104-111 (2012-08-07)
Pesticides are developed with carriers to improve their physicochemical properties and, accordingly, the bioefficacy of the applied formulation. For foliar-applied herbicide, generally less than 0.1% of the active ingredient reaching the target site could reduce pesticide performance. Recently, a carrier
Hsin-Ling Lee et al.
Clinical toxicology (Philadelphia, Pa.), 47(7), 651-658 (2009-08-12)
Most of the glyphosate-surfactant herbicides (GlySH) are formulated commercial products containing isopropylamine (IPA) salt of glyphosate (IPAG), variable amount of a surfactant, and water. Although glyphosate is only slightly toxic to rats, ingestion of GlySH may lead to severe effects
Jacek Lipok et al.
Ecotoxicology and environmental safety, 73(7), 1681-1688 (2010-09-04)
The toxicity of commercial formulation of Roundup® 360 SL, widely used, nonselective herbicide and its main constituents, glyphosate (PMG), equimolar (1:1) isopropylamine salt of glyphosate (GIPA) and isopropylamine (IPA) was examined towards eight aquatic microphotoautotrophs; seven cyanobacterial strains representing either
Katrijn De Klerck et al.
Journal of chromatography. A, 1234, 72-79 (2011-12-07)
In chiral supercritical fluid chromatography (SFC), mobile-phase additives are often used to improve enantioseparations and peak shapes. An acidic or basic additive is chosen, depending on the nature of the compound. This work highlights the simultaneous use of the acidic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service