Skip to Content
Merck
All Photos(3)

Key Documents

318191

Sigma-Aldrich

Sodium phenoxide trihydrate

98%

Synonym(s):

Sodium phenolate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€36.40
100 G
€59.40

€36.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€36.40
100 G
€59.40

About This Item

Linear Formula:
NaOC6H5 · 3H2O
CAS Number:
Molecular Weight:
170.14
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€36.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

core: sodium

mp

61-64 °C (lit.)

SMILES string

[Na+].[H]O[H].[H]O[H].[H]O[H].[O-]c1ccccc1

InChI

1S/C6H6O.Na.3H2O/c7-6-4-2-1-3-5-6;;;;/h1-5,7H;;3*1H2/q;+1;;;/p-1

InChI key

QAJGUMORHHJFNJ-UHFFFAOYSA-M

Application

<ul>
<li><strong>High-throughput quantification of ammonium content in Arabidopsis</strong>: Research utilizing sodium phenoxide trihydrate for detecting ammonium content in Arabidopsis thaliana leaves (Vega-Mas et al., 2015).</li>
<li><strong>Catalytic hydrolytic cleavage and oxy-cleavage of lignin linkages</strong>: Study on the effectiveness of sodium phenoxide trihydrate in the catalytic cleavage of lignin linkages (Xia et al., 2014).</li>
<li><strong>Ultrasonic-assisted technique as a novel method for removal of naphthenic acid from model oil using piperidinium-based ionic liquids</strong>: Investigation on the use of sodium phenoxide trihydrate in the synthesis of ionic liquids for removing naphthenic acid (Khaidzir et al., 2021).</li>
</ul>

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service