Skip to Content
Merck
All Photos(1)

Documents

309869

Sigma-Aldrich

3-Acetyl-1-methylpyrrole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.538 (lit.)

bp

148-150 °C/15 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccn(C)c1

InChI

1S/C7H9NO/c1-6(9)7-3-4-8(2)5-7/h3-5H,1-2H3

InChI key

SZYIVZGXCXFXDN-UHFFFAOYSA-N

General description

The enthalpy of formation of 3-acetyl-1-methylpyrrole in the condensed and gas phases was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Manuel A V Ribeiro da Silva et al.
The journal of physical chemistry. B, 114(8), 2846-2851 (2010-02-10)
This work reports the enthalpies of formation in the condensed and gas phases of 2-acetyl-1-methylpyrrole and 3-acetyl-1-methylpyrrole, derived from the standard (p(o) = 0.1 MPa) molar enthalpies of combustion, in oxygen, Delta(c)H(m)(o), measured by static bomb combustion calorimetry and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service