Skip to Content
Merck
All Photos(2)

Key Documents

306738

Sigma-Aldrich

1,2-Benzenedimethanethiol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2SH)2
CAS Number:
Molecular Weight:
170.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

bp

160 °C/20 mmHg (lit.)

mp

43-45 °C (lit.)

functional group

thiol

SMILES string

SCc1ccccc1CS

InChI

1S/C8H10S2/c9-5-7-3-1-2-4-8(7)6-10/h1-4,9-10H,5-6H2

InChI key

NNJWFWSBENPGEY-UHFFFAOYSA-N

Application

1,2-Benzenedimethanethiol was used to stabilize lead sulfide (PbS) quantum dots[1]. It was also used in the synthesis of 1,3-dihydrobenzo[c]thiophene[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intramolecular Condensation of 1, 2-C6H4 (CH2RH) 2 (R= O, S, and NH) to Yield Heterocyclic Compounds over Halide-cluster Catalysts.
Nagashima S, et al.
Chemistry Letters (Jpn), 40(1), 78-80 (2011)
Anirban Das et al.
Ultrasonics sonochemistry, 21(2), 892-900 (2013-10-01)
Lead sulfide (PbS) quantum dots stabilized by 1,2-benzenedimethanethiol can be synthesized by mixing Pb(NO3)2 and Na2S solutions in ethanol under ultrasound irradiation. The PbS quantum dots (2.7 and 3.6 nm in diameter) are characterized by their absorption and fluorescence spectra
Tyler Guin et al.
Nature communications, 9(1), 2531-2531 (2018-06-30)
Liquid crystalline elastomers (LCEs) are soft, anisotropic materials that exhibit large shape transformations when subjected to various stimuli. Here we demonstrate a facile approach to enhance the out-of-plane work capacity of these materials by an order of magnitude, to nearly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service