Skip to Content
Merck
All Photos(1)

Key Documents

305308

Sigma-Aldrich

N,N′-Disuccinimidyl oxalate

85%

Synonym(s):

Di(N-succinimidyl) oxalate, N,N′-Disuccinimidyl oxalate, DSO

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€131.00

€131.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€131.00

About This Item

Empirical Formula (Hill Notation):
C10H8N2O8
CAS Number:
Molecular Weight:
284.18
Beilstein:
4562450
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€131.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

85%

mp

239 °C (dec.) (lit.)

application(s)

peptide synthesis

functional group

imide

SMILES string

O=C1CCC(=O)N1OC(=O)C(=O)ON2C(=O)CCC2=O

InChI

1S/C10H8N2O8/c13-5-1-2-6(14)11(5)19-9(17)10(18)20-12-7(15)3-4-8(12)16/h1-4H2

InChI key

OMAHFYGHUQSIEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Remainder disuccinimidyl carbonate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yichao Tian et al.
Scientific reports, 5, 10582-10582 (2015-06-09)
Many remarkable properties of graphene are derived from its large energy window for Dirac-like electronic states and have been explored for applications in electronics and photonics. In addition, strong electron-phonon interaction in graphene has led to efficient photo-thermo energy conversions
Characterization of the reaction products of adult human hemoglobin and disuccinimidyloxalate.
M A Marini et al.
Biopolymers, 28(12), 2195-2200 (1989-12-01)
Bhushan J Toley et al.
The Analyst, 140(22), 7540-7549 (2015-09-24)
We present a method of rapid isothermal amplification of DNA without initial heat denaturation of the template, and methods and probes for (a) real-time fluorescence detection and (b) lateral flow detection of amplicons. Isothermal strand displacement amplification (iSDA) can achieve

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service