Skip to Content
Merck
All Photos(1)

Key Documents

302775

Sigma-Aldrich

(R)-(−)-2-Methoxy-2-phenylethanol

98%

Synonym(s):

(−)-β-Methoxyphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)CH2OH
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D −133°, c = 1 in acetone

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.52 (lit.)

bp

65 °C/0.1 mmHg (lit.)

density

1.05 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl
phenyl

SMILES string

CO[C@@H](CO)c1ccccc1

InChI

1S/C9H12O2/c1-11-9(7-10)8-5-3-2-4-6-8/h2-6,9-10H,7H2,1H3/t9-/m0/s1

InChI key

JDTUPLBMGDDPJS-VIFPVBQESA-N

Application

(R)-(-)-2-Methoxy-2-phenylethanol can be employed as a precursor to prepare:
  • (−)-3-[(2R)-2-Methoxy-2-phenylethoxy]phenol, a key intermediate for the synthesis of C4-symmetric tetraalkoxy[4]resorcinarenes using boron trifluoride catalyst.[1]
  • N-substituent of the 6,7-benzomorphan scaffold that is used as a potent opioid receptor agonist.[2]
  • Sodium methoxyphenylethoxide (Na-MPE), which is used as an oxy-initiator in the anionic polymerization of n-hexyl isocyanate.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(2S)-N-2-methoxy-2-phenylethyl-6, 7-benzomorphan compound (2S-LP2): Discovery of a biased mu/delta opioid receptor agonist
Pasquinucci L, et al.
European Journal of Medicinal Chemistry, 168(27), 189-198 (2019)
The Diastereoselective Formation of Tetraalkoxy [4] resorcinarenes Derived from (-)-(2R)-2-Methoxy-2-phenylethanol and Proof of Absolute Configurations
Bulman PPC, et al.
European Journal of Organic Chemistry, 2011(27), 5347-5354 (2011)
Effects of different reactive oxyanionic initiators on the anionic polymerizaition of n-hexyl isocyanate.
Min J, et al.
Macromolecules, 44(9), 3211-3216 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service