Skip to Content
Merck
All Photos(1)

Key Documents

301663

Sigma-Aldrich

9-Fluorenone-1-carboxylic acid

99%

Synonym(s):

9-Oxo-1-fluorenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O3
CAS Number:
Molecular Weight:
224.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

196-198 °C (lit.)

functional group

carboxylic acid
ketone

SMILES string

OC(=O)c1cccc2-c3ccccc3C(=O)c12

InChI

1S/C14H8O3/c15-13-10-5-2-1-4-8(10)9-6-3-7-11(12(9)13)14(16)17/h1-7H,(H,16,17)

InChI key

CBEFMGJHEKAMNI-UHFFFAOYSA-N

General description

9-Fluorenone-1-carboxylic acid is formed as an intermediate during four-ring polycyclic aromatic hydrocarbon fluoranthene degradation by Pseudomonas alcaligenes strain PA-10.

Application

9-Fluorenone-1-carboxylic acid was used in preparation of (E)- and (Z)-isomers of twisted 1,1′-dibromobifluorenylidene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Gordon et al.
Biodegradation, 12(6), 393-400 (2002-06-08)
Pseudomonas alcaligenes strain PA-10 degrades the four-ring polycyclic aromatic hydrocarbon fluoranthene, co-metabolically. HPLC analysis of the growth medium identified four intermediates, 9-fluorenone-1-carboxylic acid; 9-hydroxy-1-fluorene carboxylic acid; 9-fluorenone and 9-fluorenol, formed during fluoranthene degradation. Pre-exposure of PA-10 to 9-fluorenone-1-carboxylic acid and
Hindy E Bronstein et al.
Journal of the American Chemical Society, 124(30), 8870-8875 (2002-07-26)
Diindeno[1,2,3,4-defg;1',2',3',4'-mnop]chrysene (1), the smallest possible alkene-centered C60 substructure with a curved pi-system, is obtained in 25-35% yield by flash vacuum pyrolysis of the twisted 1,1'-dibromobifluorenylidene (2) on a 100 mg scale at 1050 degrees C. At 1200 degrees C, the
Jingnan Jin et al.
Environmental science and pollution research international, 24(1), 363-371 (2016-10-11)
In this study, a gram-positive fluoranthene-degrading bacterial strain was isolated from crude oil in Dagang Oilfield and identified as Microbacterium paraoxydans JPM1 by the analysis of 16S rDNA sequence. After 25 days of incubation, the strain JPM1 could degrade 91.78 % of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service