Skip to Content
Merck
All Photos(3)

Key Documents

301566

Sigma-Aldrich

2,2,3,3-Tetramethylcyclopropanecarboxylic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4C3HCO2H
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

120-121 °C (lit.)

functional group

carboxylic acid

SMILES string

CC1(C)C(C(O)=O)C1(C)C

InChI

1S/C8H14O2/c1-7(2)5(6(9)10)8(7,3)4/h5H,1-4H3,(H,9,10)

InChI key

SFHVXKNMCGSLAR-UHFFFAOYSA-N

General description

2,2,3,3-Tetramethylcyclopropanecarboxylic acid is an inactive cyclopropyl analog of valproic acid (major antiepileptic drug).[1] Inhibition of histone deacetylases and tumor cells cytotoxicities of 2,2,3,3-tetramethylcyclopropanecarboxylic acid was studied.[2]

Application

2,2,3,3-Tetramethylcyclopropanecarboxylic acid was used as a starting material for the synthesis of central nervous system-active compounds.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Eyal et al.
Biochemical pharmacology, 69(10), 1501-1508 (2005-04-29)
The tumor cells toxicity of the antiepileptic drug valproic acid (VPA) has been associated with the inhibition of histone deacetylases (HDACs). We have assessed, in comparison to VPA, the HDACs inhibition and tumor cells cytotoxicities of CNS-active VPA's constitutional isomers
Preparation of tetramethylcyclopropanecarboxylic-14C acid and insecticidal terallethrin-14C from it.
A Yoshitake et al.
Radioisotopes, 30(8), 452-454 (1981-08-01)
Neta Pessah et al.
Journal of medicinal chemistry, 52(8), 2233-2242 (2009-03-20)
2,2,3,3-Tetramethylcyclopropanecarboxylic acid (TMCA, 4) is a cyclic analogue of the antiepileptic drug (AED) valproic acid (VPA) (1). alpha-F, alpha-Cl, alpha-Br, and alpha-methyl derivatives of 4 and their amides were synthesized and tested in rodent models for anticonvulsant potency and AED-induced
Tawfeeq Shekh-Ahmad et al.
Epilepsy research, 98(2-3), 238-246 (2011-11-08)
Valproic acid (VPA) is a major antiepileptic drug (AED) that is less potent than other AEDs. 2,2,3,3-Tetramethylcyclopropanecarboxylic acid (TMCA) is an inactive cyclopropyl analogue of VPA that serves as a starting material for the synthesis of CNS-active compounds. New conjugation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service