Select a Size
About This Item
Recommended Products
Quality Level
Assay
95%
form
solid
bp
212 °C/12 mmHg (lit.)
mp
46-52 °C (lit.)
functional group
ketone
phenyl
SMILES string
CCCCCCCCCCCCCC(=O)c1ccccc1
InChI
1S/C20H32O/c1-2-3-4-5-6-7-8-9-10-11-15-18-20(21)19-16-13-12-14-17-19/h12-14,16-17H,2-11,15,18H2,1H3
InChI key
LXUIUVLDNRQBQJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service