Skip to Content
Merck
All Photos(1)

Key Documents

298417

Sigma-Aldrich

(R)-(−)-Leucinol

98%

Synonym(s):

(R)-2-Amino-4-methyl-1-pentanol, D-Leucinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€27.10
5 G
€66.60

€27.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€27.10
5 G
€66.60

About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CH2OH
CAS Number:
Molecular Weight:
117.19
Beilstein:
1719241
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€27.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

optical activity

[α]20/D −4°, c = 9 in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.4496 (lit.)

bp

198-200 °C/768 mmHg (lit.)

density

0.917 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)C[C@@H](N)CO

InChI

1S/C6H15NO/c1-5(2)3-6(7)4-8/h5-6,8H,3-4,7H2,1-2H3/t6-/m1/s1

InChI key

VPSSPAXIFBTOHY-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klaus Ringsborg Westphal et al.
Toxins, 11(5) (2019-05-19)
Fungal non-ribosomal peptide synthetase (NRPS) clusters are spread across the chromosomes, where several modifying enzyme-encoding genes typically flank one NRPS. However, a recent study showed that the octapeptide fusaoctaxin A is tandemly synthesized by two NRPSs in Fusarium graminearum. Here
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service