Skip to Content
Merck
All Photos(1)

Key Documents

298263

Sigma-Aldrich

Yttrium(III) chloride

beads, anhydrous, −10 mesh, 99.9%

Synonym(s):

Yttrium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YCl3
CAS Number:
Molecular Weight:
195.26
EC Number:
MDL number:
UNSPSC Code:
12352302

grade

anhydrous

Assay

99.9%

form

beads

impurities

<100 ppm

particle size

−10 mesh

mp

721 °C (lit.)

density

2.67 g/mL at 25 °C (lit.)

SMILES string

Cl[Y](Cl)Cl

InChI

1S/3ClH.Y/h3*1H;/q;;;+3/p-3

InChI key

PCMOZDDGXKIOLL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Will react with potassium alkoxotitanates to form precursors to novel titanium-containing ceramics

Packaging

Packaged under argon in ampules

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Watanabe et al.
Radiation protection dosimetry, 114(4), 509-513 (2005-05-19)
The efficacy of diethylenetriaminepentaacetate calcium trisodium (CaNa(3)DTPA) in a dose of 34.7 micromol kg(-1) as a function of its route of administration was investigated in rats with a puncture wound contaminated by (90)Y-chloride at a concentration of 2.55 MBq kg(-1).
S Hirano et al.
Toxicology and applied pharmacology, 121(2), 224-232 (1993-08-01)
Although radioactive yttrium (Y) has been used for medical treatment, little attention has been directed toward the toxicity of Y. We report time-course and dose-related changes in tissue distribution, subcellular localization, clearance, and acute toxicity of iv-injected yttrium chloride (YCl3)
Acceleration of the substitution of silanes with Grignard reagents by using either LiCl or YCl3/MeLi.
Naoki Hirone et al.
Angewandte Chemie (International ed. in English), 49(42), 7762-7764 (2010-09-08)
Jérôme Hannedouche et al.
Chemical communications (Cambridge, England), (30)(30), 3552-3554 (2008-07-26)
A facile method for the preparation of highly active and enantioselective yttrium precatalysts for asymmetric hydroamination of gem-disubstituted aminoalkenes, from the combination of YCl(3) or YCl(3)(THF)(3.5) with ligand (R)- and n-BuLi is described.
K Marubashi et al.
Toxicology letters, 99(1), 43-51 (1998-11-04)
We investigated pulmonary clearance of yttrium (Y) and acute lung injury following intratracheal instillation (i.t.) of yttrium chloride (YCl3) in saline- or YCl3-pretreated rats (30 days before the second challenge). About 67% of the initial dose of Y remained in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service