All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H12O4
CAS Number:
Molecular Weight:
208.21
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
mp
69-71 °C (lit.)
SMILES string
COC(=O)[C@H]1O[C@@H]1c2ccc(OC)cc2
InChI
1S/C11H12O4/c1-13-8-5-3-7(4-6-8)9-10(15-9)11(12)14-2/h3-6,9-10H,1-2H3/t9-,10+/m1/s1
InChI key
CVZUMGUZDAWOGA-ZJUUUORDSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Leukocytosis and low serum IgA in workers exposed to the epoxy compound, t-methyl-3-phenylglycidate.
Y Yamada et al.
International archives of occupational and environmental health, 60(1), 7-14 (1988-01-01)
Forty-nine out of 54 male workers engaged in the production of an epoxy compound, t-methyl-3-phenylglycidate, showed skin symptoms in varying degrees that may be due to the skin-irritative effect of the compound. The exposed workers were also shown to have
Ritsuo Imashiro et al.
The Journal of organic chemistry, 68(3), 974-979 (2003-02-01)
An efficient synthesis of methyl (2R,3S)-3-(4-methoxyphenyl)glycidate (-)-2, a key intermediate for diltiazem (1), has been developed on the basis of the highly enantioselective Mukaiyama aldol reaction of p-anisaldehyde (4a) with alpha,alpha-dichloro ketene silyl acetal 5. Thus, the reaction using a
Occupational contact dermatitis from trans-methyl-3-(4-methoxyphenyl)glycidate.
H Buisson et al.
Contact dermatitis, 25(4), 262-263 (1991-10-01)
Li Gao et al.
Journal of industrial microbiology & biotechnology, 31(11), 525-530 (2004-11-19)
Lipase production and cell growth of Serratia marcescens ECU1010 were optimized in shake flasks, with lipase production being enhanced 9.5-fold (4,780 U/l) compared with the initial activity (500 U/l). Optimal carbon and nitrogen sources were Tween-80 and peptone, and the
Dermatitis from methyl 2,3 epoxy-3-(4-methoxyphenyl)propionate.
E Rudzki et al.
Contact dermatitis, 23(5), 382-382 (1990-11-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service