Skip to Content
Merck
All Photos(1)

Key Documents

296457

Sigma-Aldrich

Octanoic acid-1-13C

99 atom % 13C

Synonym(s):

Caprylic acid-1-13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€203.00

€203.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€203.00

About This Item

Linear Formula:
CH3(CH2)613CO2H
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€203.00


Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.4284 (lit.)

bp

237 °C (lit.)

mp

16 °C (lit.)

density

0.910 g/mL at 25 °C

mass shift

M+1

SMILES string

CCCCCCC[13C](O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)/i8+1

InChI key

WWZKQHOCKIZLMA-VJJZLTLGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Octanoic-1-13C acid is an isotopic analogue of octanoic acid.

Application

Octanoic-1-13C acid is used to study the dehydrogenase of medium chain acyl-CoA using Resonance Raman (RR) study.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nishina et al.
Journal of biochemistry, 111(6), 699-706 (1992-06-01)
Resonance Raman (RR) spectra of the complex of pig kidney medium-chain acyl-CoA dehydrogenase with acetoacetyl-CoA and of the purple complex formed upon the addition of octanoyl-CoA to the dehydrogenase were obtained. RR spectra were also measured for the complexes prepared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service