Skip to Content
Merck
All Photos(1)

Key Documents

29512

Sigma-Aldrich

(R)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine]

≥97%, optical purity ee: ≥99%

Synonym(s):

(R)-3,5-t-Bu-4-MeO-MeOBIPHEP, (R)-[6,6′-Dimethoxy(1,1′-biphenyl)-2,2′-diyl]bis{bis[3,5-bis(1,1-dimethylethyl)-4-methoxyphenyl]phosphine}, SL-A109-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H104O6P2
CAS Number:
Molecular Weight:
1151.56
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

optical purity

ee: ≥99%

InChI

1S/C74H104O6P2/c1-67(2,3)49-37-45(38-50(63(49)77-27)68(4,5)6)81(46-39-51(69(7,8)9)64(78-28)52(40-46)70(10,11)12)59-35-31-33-57(75-25)61(59)62-58(76-26)34-32-36-60(62)82(47-41-53(71(13,14)15)65(79-29)54(42-47)72(16,17)18)48-43-55(73(19,20)21)66(80-30)56(44-48)74(22,23)24/h31-44H,1-30H3

InChI key

JSSIQFXOSWLAGA-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Application

Atropisomeric MeOBIPHEP ligands

  • Asymmetrical conjugate addition of arylboronic acids to maleimides and enones catalyzed by rhodium complexes
  • Intramolecular ketone hydroacylation
  • Desymmetrization through enantioselective C-C bond activation and ring expansion of allenyl cyclobutanols
  • Hydroamination of bicyclic alkenes and dienes with anilines
  • 1,4-addition/asymmetric protonation of amino acrylates

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service