Skip to Content
Merck
All Photos(1)

Key Documents

294802

Sigma-Aldrich

Chloroethane solution

2.0 M in diethyl ether, anhydrous

Synonym(s):

Ethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Cl
CAS Number:
Molecular Weight:
64.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

anhydrous

vapor pressure

32.29 psi ( 55 °C)
9.86 psi ( 20 °C)

concentration

2.0 M in diethyl ether

impurities

<0.005% water

density

0.765 g/mL at 25 °C

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

CCCl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3

InChI key

HRYZWHHZPQKTII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 1 - Repr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Banerjee et al.
Journal of dentistry, 38(6), 475-479 (2010-03-13)
Compare clinical effectiveness of sodium bicarbonate and bioactive glass powders used for dental prophylaxis. 25 patients were allocated to either good or poor oral hygiene subgroups (n=50). Using a double-blind, split-mouth model, all patients underwent prophylaxis treatment on mandibular teeth;
Lindsey L Cohen et al.
The Clinical journal of pain, 25(6), 490-494 (2009-06-23)
The purpose of this study was to evaluate the effectiveness of vapocoolant for preschoolers' immunization injection pain relief. Fifty-seven 4 to 6-year-old children were randomized into vapocoolant alone or typical care conditions. Pain was measured at the baseline and at
Andreas Tiehm et al.
Current opinion in biotechnology, 22(3), 415-421 (2011-03-08)
Because of a range of different industrial activities, sites contaminated with chloroethenes are a world-wide problem. Chloroethenes can be biodegraded by reductive dechlorination under anaerobic conditions as well as by oxidation under aerobic conditions. The tendency of chloroethenes to undergo
David J Steward
Paediatric anaesthesia, 22(3), 275-277 (2011-10-26)
Charles Robson (Figure 1) was born in New Westminster, British Columbia (now a suburb of Vancouver), in 1884 and graduated in medicine from McGill University in Montreal in 1913. Having interned and with some anesthesia training at the Royal Victoria
Robert N Richards
Journal of cutaneous medicine and surgery, 13(5), 253-256 (2009-09-23)
Botulinum toxin injections are effective in the treatment of palmar and plantar hyperhidrosis, but discomfort has limited its use. To study the use of ethyl chloride medium-stream spray in reducing injection discomfort. We used ethyl chloride medium-stream spray, in conjunction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service