Skip to Content
Merck
All Photos(1)

Documents

292613

Sigma-Aldrich

4,4′-Dianilino-1,1′-binaphthyl-5,5′-disulfonic acid dipotassium salt

Synonym(s):

Bis-ANS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5NHC10H5(SO3K)]2
CAS Number:
Molecular Weight:
636.78
Council of Europe no.:
0000
MDL number:
UNSPSC Code:
12352103

SMILES string

[K+].[K+].[O-]S(=O)(=O)c1cccc2c(ccc(Nc3ccccc3)c12)-c4ccc(Nc5ccccc5)c6c(cccc46)S([O-])(=O)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service