Skip to Content
Merck
All Photos(1)

Key Documents

292516

Sigma-Aldrich

5-Bromo-2,4-dimethoxybenzaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
245.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

140-141 °C (lit.)

SMILES string

COc1cc(OC)c(C=O)cc1Br

InChI

1S/C9H9BrO3/c1-12-8-4-9(13-2)7(10)3-6(8)5-11/h3-5H,1-2H3

InChI key

PXDIELLGFUEAIX-UHFFFAOYSA-N

General description

5-Bromo-2,4-dimethoxybenzaldehyde undergoes coupling with benzo[b]thiophene-2-boronic acid in the presence of tetrakis(triphenylphosphine)palladium(0) to give 5-(benzo[b]thien-2-yl)-2,4-dimethoxybenzaldehyde[1].

Application

5-Bromo-2,4-dimethoxybenzaldehyde has been used in the preparation of 1,2-dihydro-4,6-dimethoxy-3-methylbenzocyclobuten-1-ol[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Q Meng et al.
Bioorganic & medicinal chemistry letters, 14(6), 1513-1517 (2004-03-10)
Novel chalcone derivatives have been discovered as potent inhibitors of TNF-alpha-induced VCAM-1 expression. Thienyl or benzothienyl substitution at the meta-position of ring B helps boost potency while large substitution at the para-position on ring B is detrimental. Various substitutions are
An Efficient Synthesis of 5, 7-Dihydroxy-4-methylisobenzofuran-1 (3H)-one, a Metabolite of Aspergillus Flavus and a Key Intermediate in the Synthesis of Mycophenolic Acid.
Kobayashi K, et al.
Bulletin of the Chemical Society of Japan, 63(8), 2435-2437 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service