Skip to Content
Merck
All Photos(1)

Key Documents

291919

Sigma-Aldrich

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2 · HCl
CAS Number:
Molecular Weight:
229.70
Beilstein:
3634126
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

260-265 °C (lit.)

solubility

soluble 25 mg/mL, clear, colorless (1N NaOH in methanol)

SMILES string

Cl.COc1cc2CCNCc2cc1OC

InChI

1S/C11H15NO2.ClH/c1-13-10-5-8-3-4-12-7-9(8)6-11(10)14-2;/h5-6,12H,3-4,7H2,1-2H3;1H

InChI key

SHOWAGCIRTUYNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride is also referred as heliamine. It is an alkaloid isolated from mexican cereoid, Backebergia militaris[1].

Application

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride has been used as starting material for the synthesis of more complex isoquinolines[2] and quinolizidines[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterocycles, 34, 1857-1857 (1992)
R Mata et al.
Journal of pharmaceutical sciences, 69(1), 94-95 (1980-01-01)
Backebergia militaris (Andot) Bravo ex Sánchez Mejorada yielded alkaloid crystals from a fractionated ethanol extract of only 20 g of plant material. The alkaloid was identified as heliamine (6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline) hydrochloride. A second alkaloid, 3,4-dimethoxy-beta-phenethylamine hydrochloride, was crystallized after preparative TLC
E Glusa et al.
Die Pharmazie, 45(6), 408-410 (1990-06-01)
The synthesis of the title compounds starting from 2-Chlormethylbenzdioxan and Tetrahydroisochinolines is presented. Their actions on the platelet aggregation and the inhibition of alpha-adrenoceptors at the isolated rabbit aorta and the vas deferens of the guinea pig were investigated.
S A Barker et al.
Biochemical pharmacology, 30(17), 2461-2468 (1981-09-01)
Gas chromatographic/mass spectrometric data are presented which demonstrate the presence of 6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline (DHTIQ) as a normal constituent of rat brain. The level of DHTIQ was calculated to be 10.0 +/- 3.0 ng/g wet weight (+/- S.D., N = 9) of
Ken-ichi Umehara et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(7), 1427-1433 (2009-04-11)
(-)-N-{2-[(R)-3-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-carbonyl)piperidino]ethyl}-4-fluorobenzamide (YM758), a novel "funny" If current channel (If channel) inhibitor, is developed as a treatment for stable angina and atrial fibrillation. In this study, the pharmacokinetic/pharmacodynamic (PK/PD) relationship after intravenous administration of YM758 to tachycardia-induced dogs was investigated and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service