Skip to Content
Merck
All Photos(1)

Key Documents

29067

Diiodo(p-cymene)ruthenium(II) dimer

Synonym(s):

Di-μ-iodo(p-cymene)iodoruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28I4Ru2
CAS Number:
Molecular Weight:
978.19
MDL number:

mp

234-236 °C

SMILES string

I[Ru]1[I][Ru](I)[I]1.C[C]2[CH][CH][C]([CH][CH]2)C(C)C.C[C]3[CH][CH][C]([CH][CH]3)C(C)C

InChI

1S/2C10H14.2HI.2I.2Ru/c2*1-8(2)10-6-4-9(3)5-7-10;;;;;;/h2*4-8H,1-3H3;2*1H;;;;/q;;;;;;2*+1/p-2

InChI key

GMEPSZJJBHGIDB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst in:
  • Ruthenium-catalyzed hydrogenation
  • Preparation of Walphos-type ferrocenyl-aryl diphosphine ligands
  • Ruthenium-catalyzed synthesis reactions
  • Enantioselective hydrogenation of ketones and olefins

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service