Skip to Content
Merck
All Photos(1)

Key Documents

290394

Sigma-Aldrich

2,5-Difluorophenol

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€30.50
5 G
€62.50

€30.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€30.50
5 G
€62.50

About This Item

Linear Formula:
F2C6H3OH
CAS Number:
Molecular Weight:
130.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€30.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

40-42 °C (lit.)

functional group

fluoro

SMILES string

Oc1cc(F)ccc1F

InChI

1S/C6H4F2O/c7-4-1-2-5(8)6(9)3-4/h1-3,9H

InChI key

INXKVYFOWNAVMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Conversion of 2,5-difluorophenol by whole cells of Rhodococcus opacus 1G has been investigated by 9F NMR analysis[1].

Application

2,5-Difluorophenol has been used in the synthesis of di- or trifluorinated hydroxybenzoic acids[2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The regioexhaustive functionalization of difluorophenols and trifluorophenols through organometallic intermediates.
Marzi E, et al.
Synthesis, 2004(10), 1609-1618 (2004)
V S Bondar et al.
FEMS microbiology letters, 181(1), 73-82 (1999-11-24)
The regiospecificity of hydroxylation of C2-halogenated phenols by Rhodococcus opacus 1G was investigated. Oxidative defluorination at the C2 position ortho with respect to the hydroxyl moiety was preferred over hydroxylation at the non-fluorinated C6 position for all 2-fluorophenol compounds studied.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service