Skip to Content
Merck
All Photos(1)

Key Documents

290254

Sigma-Aldrich

3,5-Difluorobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€61.70
25 G
€179.00

€61.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€61.70
25 G
€179.00

About This Item

Linear Formula:
F2C6H3COCl
CAS Number:
Molecular Weight:
176.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€61.70


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.5031 (lit.)

functional group

acyl chloride
fluoro

SMILES string

Fc1cc(F)cc(c1)C(Cl)=O

InChI

1S/C7H3ClF2O/c8-7(11)4-1-5(9)3-6(10)2-4/h1-3H

InChI key

OYZWEOORLJBPMA-UHFFFAOYSA-N

Application

3,5-Difluorobenzoyl chloride was used in the synthesis of AMPA (α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid) potentiator LY450108-[14C] containing 14C-label attached to the chiral center of the molecule[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 14C?labeled and tritiated AMPA potentiator LY450108.
Journal of Labelled Compounds & Radiopharmaceuticals, 48(2), 85-100 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service