Skip to Content
Merck
All Photos(1)

Key Documents

290084

Sigma-Aldrich

2-Amino-4-methyl-3-nitropyridine

98%

Synonym(s):

2-Amino-3-nitro-4-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O2
CAS Number:
Molecular Weight:
153.14
Beilstein:
139111
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

136-140 °C (lit.)

SMILES string

Cc1ccnc(N)c1[N+]([O-])=O

InChI

1S/C6H7N3O2/c1-4-2-3-8-6(7)5(4)9(10)11/h2-3H,1H3,(H2,7,8)

InChI key

IKMZGACFMXZAAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The crystal structure of 2-amino-4-methyl-3-nitropyridine was elucidated.

Application

2-Amino-4-methyl-3-nitropyridine was used in the synthesis of 2-amino-5-hydroxy-4-methyl-3-nitropyridine, 2-amino-4-hydroxymethyl-3-nitropyridine and 2-amino-4-methyl-3-nitropyridine-1-oxide by undergoing biotransformation by Cunninghamella elegans ATCC 26269.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Tully et al.
Journal of industrial microbiology & biotechnology, 39(12), 1789-1799 (2012-08-29)
Biotransformation of the highly substituted pyridine derivative 2-amino-4-methyl-3-nitropyridine by Cunninghamella elegans ATCC 26269 yielded three products each with a molecular weight of 169 Da which were identified as 2-amino-5-hydroxy-4-methyl-3-nitropyridine, 2-amino-4-hydroxymethyl-3-nitropyridine, and 2-amino-4-methyl-3-nitropyridine-1-oxide. Biotransformation by Streptomyces antibioticus ATCC 14890 gave two different
I Bryndal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 96, 952-962 (2012-09-04)
The crystal structures of 2-amino-4-methyl-3-nitropyridine (I), 2-amino-4-methyl-3,5-dinitropyridine (II) and 2-amino-4-methyl-5-nitropyridine (III) have been determined. The compounds crystallize in the monoclinic P2(1)/n, triclinic P-1 and monoclinic C2/c space groups, respectively. These structures are stabilized by a combination of N-H···N and N-H···O

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service