Skip to Content
Merck
All Photos(2)

Key Documents

288063

Sigma-Aldrich

Tetrakis(triphenylphosphite)nickel(0)

Synonym(s):

Nickel tetrakis(triphenoxyphosphine), Tetrakis(triphenoxyphosphine)nickel, [(PhO)3P]4Ni

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€69.30

€69.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€69.30

About This Item

Linear Formula:
[(C6H5O)3P]4Ni
CAS Number:
Molecular Weight:
1303.86
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€69.30


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

reaction suitability

core: nickel
reagent type: catalyst

mp

145 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Ni].O(P(Oc1ccccc1)Oc2ccccc2)c3ccccc3.O(P(Oc4ccccc4)Oc5ccccc5)c6ccccc6.O(P(Oc7ccccc7)Oc8ccccc8)c9ccccc9.O(P(Oc%10ccccc%10)Oc%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15O3P.Ni/c4*1-4-10-16(11-5-1)19-22(20-17-12-6-2-7-13-17)21-18-14-8-3-9-15-18;/h4*1-15H;

InChI key

PGXCIAAANFTEKX-UHFFFAOYSA-N

Application

Catalyst for alkene and alkyne hydrocyanation and styrenedimerization

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fitzmaurice, N. J.; Jackson, W. R. et al.
Journal of Organometallic Chemistry, 285, 375-375 (1985)
Andell, O. S.; Baeckvall, J. E. et al.
Acta Chemica Scandinavica, B40, 184-184 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service