Skip to Content
Merck
All Photos(1)

Key Documents

285781

Sigma-Aldrich

β-Estradiol 3-benzoate

98%

Synonym(s):

1,3,5(10)-Estratriene-3,17β-diol 3-benzoate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-benzoate, 3-Benzoyloxy-17β-estrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28O3
CAS Number:
Molecular Weight:
376.49
Beilstein:
3107526
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

optical activity

[α]25/D +60°, c = 1 in dioxane

mp

191-198 °C (lit.)

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])CCc4cc(OC(=O)c5ccccc5)ccc24

InChI

1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1

InChI key

UYIFTLBWAOGQBI-BZDYCCQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Martins et al.
Theriogenology, 78(5), 974-980 (2012-07-24)
Two experiments evaluated the effects of timing of the induction of ovulation in superstimulated lactating Holstein donor cows that were fixed-time artificially inseminated. Secondary objectives were to evaluate the effects of the timing of progesterone (P4) device removal (Experiment 1)
B Ariyadi et al.
Poultry science, 92(12), 3205-3213 (2013-11-16)
Mucins play an essential role as mucosal barrier to prevent invasion of pathogens in the oviductal tissue of hens. The aim of this study was to determine the effect of estradiol and lipopolysaccharide (LPS) on the mucin expression in the
Matthew Mahavongtrakul et al.
Endocrinology, 154(9), 3251-3260 (2013-07-05)
Sexual receptivity in the female rat is dependent on dose and duration of estradiol exposure. A 2 μg dose of estradiol benzoate (EB) primes reproductive behavior circuits without facilitating lordosis. However, 50 μg EB facilitates lordosis after 48 hours. Both
S A A Edwards et al.
Animal reproduction science, 137(3-4), 129-136 (2013-02-05)
The aim of this study was to investigate the effects on follicle stimulating hormone (FSH) secretion and dominant follicle (DF) growth, of treatment of Bos indicus heifers with different combinations of intra-vaginal progesterone releasing devices (IPRD), oestradiol benzoate (ODB), PGF2α
S A A Butler et al.
Reproduction in domestic animals = Zuchthygiene, 47(3), 463-471 (2012-04-14)
The primary objective of this study was to investigate the impact of animal-level factors including energy balance and environmental/management stress, on the ovarian function of Bos indicus heifers treated to synchronize ovulation. Two-year-old Brahman (BN) (n = 30) and BN-cross

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service