285773
α-Estradiol
98%
Synonym(s):
1,3,5(10)-Estratriene-3,17α-diol, 3,17α-Dihydroxy-1,3,5(10)-estratriene, Epiestradiol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
98%
impurities
<3% water
SMILES string
[H][C@]12CC[C@]3(C)[C@H](O)CC[C@@]3([H])[C@]1([H])CCc4cc(O)ccc24
InChI
1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17-,18+/m1/s1
InChI key
VOXZDWNPVJITMN-SFFUCWETSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Estradiol (known as alpha Estradiol or 17 alpha Estradiol) is a biologically active estrogen in human breast cancer cells in tissue culture.17-estradiol and its selective receptor, ER-X, are not part of a classical hormone/receptor endocrine system but of a system with important autocrine/paracrine functions in the developing and adult brain. 17-Estradiol may have enormous implications for hormone replacement strategies at the menopause and in the treatment of such neurodegenerative disorders as Alzheimer′s disease and ischemic stroke.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C Y Cheng et al.
Fertility and sterility, 32(5), 566-570 (1979-11-01)
In modified Tyrode's solution, 17 beta-estradiol at concentrations between 0.1 microgram/ml and 320 nmoles/ml was effective in increasing human spermatozoal forward migration. 17 alpha-Estradiol, although structurally similar to 17 beta-estradiol, had no effect on human spermatozoal motility. DL-Norgestrel at concentrations
J Pitt et al.
Journal of inherited metabolic disease, 25(2), 83-88 (2002-07-18)
Three patients with ketosis had increased excretion of 3-hydroxyglutarate (21.8-37.9 micromol/mmol creatinine; controls 2.3 +/- 1.6), an indicator of glutaryl-CoA dehydrogenase deficiency (GDHD), which normalized when the patients were nonketotic. Clinical assessment of all three patients and enzyme studies in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service