Skip to Content
Merck
All Photos(1)

Key Documents

28255

Sigma-Aldrich

(+)-Cuparene

≥99.0% (sum of enantiomers, GC)

Synonym(s):

(R)-1-(p-Tolyl)-1,2,2-trimethylcyclopentane

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€576.00

€576.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€576.00

About This Item

Empirical Formula (Hill Notation):
C15H22
CAS Number:
Molecular Weight:
202.34
Beilstein:
2326949
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

€576.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D +64±2°, neat

refractive index

n20/D 1.523

bp

275 °C (lit.)

density

0.937 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)[C@]2(C)CCCC2(C)C

InChI

1S/C15H22/c1-12-6-8-13(9-7-12)15(4)11-5-10-14(15,2)3/h6-9H,5,10-11H2,1-4H3/t15-/m0/s1

InChI key

SLKPBCXNFNIJSV-HNNXBMFYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Peñuelas et al.
The Plant cell, 31(10), 2491-2509 (2019-08-09)
The lipid-derived phytohormone jasmonoyl-isoleucine regulates plant immunity, growth and development in vascular plants by activating genome-wide transcriptional reprogramming. In Arabidopsis (Arabidopsis thaliana), this process is largely orchestrated by the master regulator MYC2 and related transcription factors (TFs). However, the TFs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service