Skip to Content
Merck
All Photos(1)

Key Documents

280208

Sigma-Aldrich

Dimethyl acetylsuccinate

Synonym(s):

2-Acetylbutanedioic acid-1,4-dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CCH2CH(COCH3)CO2CH3
CAS Number:
Molecular Weight:
188.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

bp

129-134 °C/12 mmHg (lit.)

mp

33 °C (lit.)

solubility

water: soluble 56.2 g/L at 20 °C

density

1.16 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

COC(=O)CC(C(C)=O)C(=O)OC

InChI

1S/C8H12O5/c1-5(9)6(8(11)13-3)4-7(10)12-2/h6H,4H2,1-3H3

InChI key

XREKLQOUFWBSFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl acetylsuccinate undergoes sulfuric acid catalyzed Pechmann condensation with the corresponding resorcinol derivative to give 7-hydroxycoumarins.

Application

Dimethyl acetylsuccinate has been used in the synthesis of 2-oxofuro[2,3-b]pyrroles and 19-methyl-15-oxa-20-azatricyclo[12.3.3.0(1,14)]icos-18-en-18-carboxylates.
Reactant involved in:
  • Cyclization for the synthesis of β-carbolines
  • Synthesis of photochromic molecules with improved fatigue resistances and dark form stabilities
  • Asymmetric hydrogenation of acylsuccinate followed by lactonization for synthesis of oxotetrahydrofurancarboxylates
  • Amidation for synthesis of amino-acid derivatives of dihydropyranocoumarins

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orazio A Attanasi et al.
The Journal of organic chemistry, 69(8), 2686-2692 (2004-04-13)
New and interesting 2-oxofuro[2,3-b]pyrroles and 19-methyl-15-oxa-20-azatricyclo[12.3.3.0(1,14)]icos-18-en-18-carboxylates have been obtained in good yields by the one-pot reaction, in basic medium, of 1,2-diaza-1,3-butadienes with diethyl or dimethyl acetylsuccinate or methyl 2-(1,3-dioxo-2-cyclotetradecyl)acetate, respectively, under mild conditions. Treatment of the same starting materials with
Raphael M Franzini et al.
Chembiochem : a European journal of chemical biology, 9(18), 2981-2988 (2008-11-28)
Templated nucleic acid detection is an emerging bioanalytical method that makes use of the target DNA or RNA strand to initiate a fluorogenic reaction. The Staudinger reduction holds particular promise for templated sensing of nucleic acids because the involved functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service