Skip to Content
Merck
All Photos(1)

Key Documents

279870

Sigma-Aldrich

(R)-(−)-2-Phenylbutyric acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(C6H5)CO2H
CAS Number:
Molecular Weight:
164.20
Beilstein:
2045208
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Assay

99%

optical activity

[α]20/D −93°, c = 0.9 in toluene

bp

102-104 °C/0.4 mmHg (lit.)

density

1.055 g/mL at 25 °C (lit.)

functional group

carboxylic acid
phenyl

SMILES string

CC[C@@H](C(O)=O)c1ccccc1

InChI

1S/C10H12O2/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,11,12)/t9-/m1/s1

InChI key

OFJWFSNDPCAWDK-SECBINFHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service