Skip to Content
Merck
All Photos(2)

Key Documents

278440

Sigma-Aldrich

2-{[2-(Dimethylamino)ethyl]methylamino}ethanol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€58.50
100 G
€133.00

€58.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€58.50
100 G
€133.00

About This Item

Linear Formula:
(CH3)2NCH2CH2N(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
146.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€58.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.4539 (lit.)

bp

207 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CN(C)CCN(C)CCO

InChI

1S/C7H18N2O/c1-8(2)4-5-9(3)6-7-10/h10H,4-7H2,1-3H3

InChI key

LSYBWANTZYUTGJ-UHFFFAOYSA-N

Application

2-{[2-(Dimethylamino)ethyl]methylamino}ethanol (dmemH) has been used in the preparation of new iron clusters:
  • [Fe7O4(O2CPh)11(dmem)2][1]
  • [Fe7O4 (O2CMe)11(dmem)2][1]
  • [Fe6O2(OH)4(O2CBut)8(dmem)2][1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rashmi Bagai et al.
Inorganic chemistry, 46(11), 4535-4547 (2007-04-26)
The syntheses, crystal structures, and magnetochemical characterization of four new iron clusters [Fe7O4(O2CPh)11(dmem)2] (1), [Fe7O4(O2CMe)11(dmem)2] (2), [Fe6O2(OH)4(O2CBut)8(dmem)2] (3), and [Fe3O(O2CBut)2(N3)3(dmem)2] (4) (dmemH=Me2NCH2CH2N(Me)CH2CH2OH)=2-{[2-(dimethylamino)ethyl]methylamino}ethanol) are reported. The reaction of dmemH with [Fe3O(O2CR)6(H2O)3](NO3) (R=Ph (1), Me (2), and But (3)) gave 1, 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service