Skip to Content
Merck
All Photos(1)

Key Documents

278033

Sigma-Aldrich

Diethyl isopropylidenemalonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=C(COOC2H5)2
CAS Number:
Molecular Weight:
200.23
Beilstein:
1776122
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.451 (lit.)

bp

175-178 °C/120 mmHg (lit.)

density

1.021 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)\C(=C(/C)C)C(=O)OCC

InChI

1S/C10H16O4/c1-5-13-9(11)8(7(3)4)10(12)14-6-2/h5-6H2,1-4H3

InChI key

WEISAZNMMVPNTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reduction of diethyl isopropylidenemalonate with sodium borohydride has been reported[1].

Application

Diethyl isopropylidenemalonate has been used in the synthesis of retinoids having functional groups at the side-chain terminus[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduction of alkylidenecyanoacetates with sodium borohydride.
Marshall JA and Carroll RD.
The Journal of Organic Chemistry, 30(8), 2748-2754 (1965)
Y F Shealy et al.
Journal of medicinal chemistry, 31(6), 1124-1130 (1988-06-01)
Retinoids that have two functional groups at the side-chain terminus have been synthesized. The two terminal functional groups are combinations of the carboxyl, carbethoxy, and N-(ethylamino)carbonyl groups. The synthesis route is based on the sodium amide catalyzed condensation of (E,E)-beta-ionylideneacetaldehyde

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service