Skip to Content
Merck
All Photos(1)

Key Documents

277444

Supelco

Jasmone

90%

Synonym(s):

cis-3-Methyl-2-(2-pentenyl)-2-cyclopenten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Beilstein:
1907713
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

Assay

90%

refractive index

n20/D 1.498 (lit.)

bp

134-135 °C/12 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

application(s)

agriculture

SMILES string

[H]\C(CC)=C(/[H])CC1=C(C)CCC1=O

InChI

1S/C11H16O/c1-3-4-5-6-10-9(2)7-8-11(10)12/h4-5H,3,6-8H2,1-2H3/b5-4-

InChI key

XMLSXPIVAXONDL-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Remainder trans isomer

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

John A Pickett et al.
Phytochemistry, 68(22-24), 2937-2945 (2007-11-21)
The challenges and opportunities for protecting agricultural production of food and other materials will be met through exploiting the induction of defence pathways in plants to control pests, diseases and weeds. These approaches will involve processes that can be activated
Eliezer Flescher
Anti-cancer drugs, 16(9), 911-916 (2005-09-16)
Since salicylate, a plant stress hormone, suppresses the growth of various types of cancer cells, it was deemed of interest to investigate whether the jasmonate family of plant stress hormones is endowed with anti-cancer activities. Cell lines representing a wide
Toby J A Bruce et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(12), 4553-4558 (2008-03-22)
It is of adaptive value for a plant to prepare its defenses when a threat is detected, and certain plant volatiles associated with insect damage, such as cis-jasmone (CJ), are known to switch-on defense metabolism. We used aphid and aphid
Walter P Suza et al.
Planta, 227(6), 1221-1232 (2008-02-06)
The Arabidopsis thaliana (L.) Heynh. JASMONATE RESISTANT 1( JAR1) locus is essential for pathogen defense, but its role in wound response has not been investigated. JAR1 encodes an enzyme that conjugates jasmonic acid (JA) to isoleucine, which was recently shown
Tomonori Misaki et al.
Journal of the American Chemical Society, 127(9), 2854-2855 (2005-03-03)
Ti-crossed-Claisen condensation between a 1:1 mixture of carboxylic esters and acid chlorides promoted by TiCl4-Bu3N-N-methylimidazole proceeded successfully to give various beta-keto esters in good yields with excellent selectivities (19 examples, approximately 48-95% yield; cross/self-selectivity = approximately 96/4-99/1). The present method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service