Skip to Content
Merck
All Photos(1)

Key Documents

275069

Sigma-Aldrich

Ethynyltributylstannane

95%

Synonym(s):

Tributylstannylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CSn(CH2CH2CH2CH3)3
CAS Number:
Molecular Weight:
315.08
Beilstein:
3537659
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.476 (lit.)

bp

70 °C/0.2 mmHg (lit.)

density

1.089 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)C#C

InChI

1S/3C4H9.C2H.Sn/c3*1-3-4-2;1-2;/h3*1,3-4H2,2H3;1H;

InChI key

YEMJHNYABQHWHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethynyltributylstannane can be used as a reactant to prepare:
  • Terminal arylacetylene derivatives by Stille coupling reaction with aryl halides in the presence of palladium catalyst.[1]
  • 1,4-bis(tributylstannyl)but-1-en-3-yne by dimerization using a metal complex having bulky ligand catalyst system.[2]
  • Isoquinolone derivatives by reacting with benzotriazinones via denitrogenative insertion in the presence of nickel catalyst.[3]
  • Enyne key intermediates in the total synthesis of (-)-acutumine and (-)-dechloroacutumine.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Total Syntheses of (-)-Acutumine and (-)-Dechloroacutumine
King SM, et al.
Angewandte Chemie (International ed. in English), 125(13), 3730-3733 (2013)
Journal of the Chemical Society. Perkin Transactions 1, 1657-1657 (1993)
Synthesis of terminal arylacetylenes by a Stille coupling reaction catalysed by a MCM-41-supported bidentate phosphine palladium (O) complex
H Wenyan, et al.
J. Chem. Res. (M), 2008(11), 615-618 (2008)
Acta Chemica Scandinavica, 48, 61-61 (1994)
Synlett, 557-557 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service