Skip to Content
Merck
All Photos(1)

Key Documents

274860

Sigma-Aldrich

4-(Diethoxymethyl)benzaldehyde

97%

Synonym(s):

Terephthalaldehyde diethyl acetal

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€57.60
25 G
€196.00

€57.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€57.60
25 G
€196.00

About This Item

Linear Formula:
(C2H5O)2CHC6H4CHO
CAS Number:
Molecular Weight:
208.25
Beilstein:
4293872
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.506 (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

aldehyde
ether

SMILES string

[H]C(=O)c1ccc(cc1)C(OCC)OCC

InChI

1S/C12H16O3/c1-3-14-12(15-4-2)11-7-5-10(9-13)6-8-11/h5-9,12H,3-4H2,1-2H3

InChI key

HTMXMFARWHNJDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshitsugu Akiyama et al.
Bioconjugate chemistry, 15(2), 424-427 (2004-03-18)
New heterotelechelic PEG-containing benzaldehyde and 2-pyridyldithio endgroup (CHO-Bz-PEG-SSpyl) was synthesized with high efficiency and high selectivity. An alpha-benzylacetal-omega-methansulfonyl PEG was prepared as the first step to CHO-Bz-PEG-SSpyl through the ring-opening polymerization of ethylene oxide (EO) initiated by potassium 4-(diethoxymethyl)benzyl alkoxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service