Skip to Content
Merck
All Photos(3)

Documents

274712

Sigma-Aldrich

N,N′-Bis(salicylidene)ethylenediaminocobalt(II)

99%

Synonym(s):

Co(salen), N,N′-Disalicylidene-ethylenediamine cobalt(II) salt, Co(SALEN)2, Ethylenebis(salicylimine) cobalt(II) salt, Salcomine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2N=CHC6H4-2-(O-)]2Co
CAS Number:
Molecular Weight:
325.23
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

reaction suitability

core: cobalt
reagent type: catalyst

SMILES string

C1C\N=C\c2ccccc2O[Co]Oc3ccccc3\C=N\1

InChI

1S/C16H16N2O2.Co/c19-15-7-3-1-5-13(15)11-17-9-10-18-12-14-6-2-4-8-16(14)20;/h1-8,11-12,19-20H,9-10H2;/q;+2/p-2/b17-11+,18-12+;

InChI key

NPAQSKHBTMUERN-OYJDLGDISA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Watanabe et al.
Antiviral chemistry & chemotherapy, 9(3), 269-274 (1999-01-06)
Salcomine, N,N'-bis(salicylidene)ethylene diamino-cobalt (II), and its derivatives were evaluated for their ability to inhibit selectively human cytomegalovirus (HCMV) proteinase activity. The 50% inhibitory concentration (IC50) of salcomine was 1.4 microM for HCMV proteinase, but > 200 microM for three other
K Otsuji et al.
Advances in space research : the official journal of the Committee on Space Research (COSPAR), 7(4), 69-72 (1987-01-01)
Oxygen concentration and separation is an essential factor for air recycling in a CELSS. Furthermore, if the value of the plant assimilatory quotient is not coincident with that of the animal respiratory quotient, the recovery of O2 from the concentrated
Adushan Pillay et al.
Organic & biomolecular chemistry, 10(38), 7809-7819 (2012-08-24)
The synthesis of two closely related pyranonaphthoquinones, dehydroherbarin and anhydrofusarubin, is described. The construction of the naphthalene nuclei was achieved using the Stobbe condensation reaction using 2,4-dimethoxybenzaldehyde and 2,4,5-trimethoxybenzaldehyde as their respective starting materials. Two key steps en route include

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service