Skip to Content
Merck
All Photos(3)

Key Documents

269999

Sigma-Aldrich

Tris(2,3-dibromopropyl) isocyanurate

97%

Synonym(s):

Hexahydro-1,3,5-tris(2,3-dibromopropyl)-1,3,5-triazine-2,4,6-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15Br6N3O3
CAS Number:
Molecular Weight:
728.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

solubility

chloroform: soluble 25 mg/mL, clear, colorless to yellow

functional group

bromo

SMILES string

BrCC(Br)CN1C(=O)N(CC(Br)CBr)C(=O)N(CC(Br)CBr)C1=O

InChI

1S/C12H15Br6N3O3/c13-1-7(16)4-19-10(22)20(5-8(17)2-14)12(24)21(11(19)23)6-9(18)3-15/h7-9H,1-6H2

InChI key

NZUPFZNVGSWLQC-UHFFFAOYSA-N

General description

Tris(2,3-dibromopropyl) isocyanurate is a non degradable heterocyclic hexabrominated flame retardant. It inhibited the expression of psaA and psbC that led to the decrease of Nannochloropsis sp.′s growth biomass.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Ruan et al.
Environmental science & technology, 43(9), 3080-3086 (2009-06-19)
A brominated flame retardant (BFR), hexabrominated heterocyclic tris-(2,3-dibromopropyl) isocyanurate (TBC), was identified,forthe first time, in the natural environment.The chemical was found in river water (2.33-163 ng/L), surface sediments (85.0 ng/g-6.03 microg/g dry weight (dw)), soils (19.6-672 ng/g dw), earthworm (9.75-78.8
Ling Wang et al.
Gene, 540(1), 68-70 (2014-03-01)
The proteins encoded by psaA and psaB form a heterodimer, an essential compound of photosystem; while the protein encoded by psbC binds with chlorophyll a in photosystem II, serving as antennae in photosystem. Here we report that a heterocyclic brominated
Peng Zhao et al.
The Analyst, 136(9), 1952-1956 (2011-03-05)
Tris(2,3-dibromopropyl) isocyanurate (TBC) is a heterocyclic hexabrominated flame retardant, which cannot be degraded even over a long time and may be a potential environmental pollutant. In this paper, TBC is for the first time as far as we know determined
Jiayong Feng et al.
Talanta, 82(5), 1929-1934 (2010-09-30)
Hexabromocyclododecanes (HBCDs) and tris (2,3-dibromopropyl) isocyanurate (TBC) are both brominated flame retardants, and have been used throughout the world with subsequent concern for potential environment pollution. Here we describe the development of a method for the simultaneous determination of TBC
Nali Zhu et al.
Environmental science & technology, 46(13), 7174-7181 (2012-06-06)
A novel brominated flame retardant (BFR), tris(2,3-dibromopropyl) isocyanurate (TBC), as well as hexabromocyclododecanes (HBCDs) and polybrominated diphenyl ethers (PBDEs), were analyzed in 11 species of mollusks collected from nine coastal cities around the Chinese Bohai Sea in 2009 and 2010.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service