Skip to Content
Merck
All Photos(1)

Key Documents

266698

Sigma-Aldrich

Cobalt

foil, thickness 1.0 mm, ≥99.99%

Synonym(s):

Cobalt element, Cobalt-59

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.99%

form

foil

resistivity

6.24 μΩ-cm, 20°C

thickness

1.0 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Kurahashi et al.
Inorganic chemistry, 52(7), 3908-3919 (2013-03-23)
The Co(salen)(X) complex, where salen is chiral N,N'-bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine and X is an external axial ligand, has been widely utilized as a versatile catalyst. The Co(salen)(X) complex is a stable solid that has been conventionally described as a Co(III)(salen)(X) complex. Recent
H John Cooper et al.
The Journal of bone and joint surgery. American volume, 95(10), 865-872 (2013-05-17)
Femoral stems with dual-taper modularity were introduced to allow additional options for hip-center restoration independent of femoral fixation in total hip arthroplasty. Despite the increasing availability and use of these femoral stems, concerns exist about potential complications arising from the
D W Su et al.
Journal of nanoscience and nanotechnology, 13(5), 3354-3359 (2013-07-19)
Highly ordered mesoporous Co3O4 nanostructures were prepared using SBA-15 silica as hard templates. The mesoporous structures were characterized by X-ray diffraction, high resolution transmission electron microscopy, and N2 adsorption/desorption isotherm analysis. The results demonstrated that the as-prepared mesoporous Co3O4 has
Pushya A Potnis et al.
Cellular immunology, 282(1), 53-65 (2013-05-18)
Metal orthopedic implant debris-induced osteolysis of hip bone is a major problem in patients with prosthetic-hips. Although macrophages are the principal targets for implant-wear debris, the receptor(s) and mechanisms underlying these responses are not fully elucidated. We examined whether the
M K Shobana et al.
Journal of nanoscience and nanotechnology, 13(5), 3535-3538 (2013-07-19)
Ferrites are extremely important magnetic ceramics in the production of electronic components because they reduce the energy losses by the induced currents acting as electrical insulators. Similarly, the spinel-structured cobalt-based ferrites are promising materials for stress, torsion sensors and energy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service