Skip to Content
Merck
All Photos(2)

Documents

264466

Sigma-Aldrich

2,6-Difluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3OH
CAS Number:
Molecular Weight:
130.09
Beilstein:
2043613
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

storage condition

protect from light

bp

59-61 °C/17 mmHg (lit.)

mp

38-41 °C (lit.)

solubility

ethanol: soluble 50 mg/mL, clear, colorless to light yellow

storage temp.

2-8°C

SMILES string

Oc1c(F)cccc1F

InChI

1S/C6H4F2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H

InChI key

CKKOVFGIBXCEIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-difluorophenol undergoes oxidative polymerization in the presence of the Fe-N,N′-bis(salicylidene)ethylenediamine (salen) complex (catalyst) and hydrogen peroxide (oxidizing agent) to give poly(2,6-difluoro-1,4-phenylene oxide).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Qiu et al.
Journal of medicinal chemistry, 42(2), 329-332 (1999-02-02)
3-(Aminomethyl)-2,6-difluorophenol (6) and 4-(aminomethyl)-2, 6-difluorophenol (7) were synthesized in eight and four steps, respectively, starting from 2,6-difluorophenol, to test the potential of the 2,6-difluorophenol moiety to act as a lipophilic bioisostere of a carboxylic acid. Compounds 6 and 7 are
Oxidative Polymerization of 2,6-Difluorophenol to Crystalline Poly(2,6-difluoro-1,4-phenylene oxide).
Ikeda R, et al.
Macromolecules, 33(18), 6648-6652 (2000)
Jiasong Li et al.
Biochemistry, 58(17), 2218-2227 (2019-04-05)
Cysteine dioxygenase (CDO) is a nonheme iron enzyme that adds two oxygen atoms from dioxygen to the sulfur atom of l-cysteine. Adjacent to the iron site of mammalian CDO, there is a post-translationally generated Cys-Tyr cofactor, whose presence substantially enhances
Akira Ohashi et al.
Talanta, 146, 789-794 (2015-12-24)
The solubilities of several cobalt(III) chelates (CoL3) with in supercritical carbon dioxide (SC-CO2) were investigated in the presence of fluorine- and trifluoromethyl-substituted phenols (PhOH) using UV-vis spectrophotometry. Solubility enhancement of CoL3 complexes in SC-CO2 was accomplished by adding PhOH. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service