All Photos(1)
About This Item
Linear Formula:
F2C6H3CH2NH2
CAS Number:
Molecular Weight:
143.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.495 (lit.)
density
1.222 g/mL at 25 °C (lit.)
SMILES string
NCc1cc(F)ccc1F
InChI
1S/C7H7F2N/c8-6-1-2-7(9)5(3-6)4-10/h1-3H,4,10H2
InChI key
GDFBHCMFIUBEQT-UHFFFAOYSA-N
Application
2,5-Difluorobenzylamine has been used as amine building block in tandem Ugi/Diels-Alder reaction on soluble polymer support.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
168.8 °F - closed cup
Flash Point(C)
76 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Parallel synthesis of tandem Ugi/Diels-Alder reaction products on a soluble polymer support directed toward split-pool realization of a small molecule library.
Oikawa M, et al.
Tetrahedron Letters, 46(3), 415-418 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service