264164
2-Chloro-9,10-bis(phenylethynyl)anthracene
Synonym(s):
2-Chloro-BPEA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C30H17Cl
CAS Number:
Molecular Weight:
412.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
mp
224-226 °C (lit.)
SMILES string
Clc1ccc2c(C#Cc3ccccc3)c4ccccc4c(C#Cc5ccccc5)c2c1
Application
Reagent for chemiluminescence research.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aldrichimica Acta, 16, 59-59 (1983)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service