Skip to Content
Merck
All Photos(1)

Key Documents

262838

Sigma-Aldrich

Tantalum

powder, ≥99.9%, −40-+200 mesh

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9%

form

powder

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

particle size

−40-+200 mesh

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Application

Employed in the synthesis of metal-rich layered tellurides, TaNi2Te2 and TaCo2Te2, which possess ideal characteristics for photovoltaic applications, for surface scanning studies, and which may give clean and atomically flat surfaces. Starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ogliaro, F. et al.
Inorganic Chemistry, 37, 6199-6199 (1998)
Neuhasen, J. et al.
Chemistry of Materials, 10, 3870-3870 (1998)
A M Ashmore et al.
Journal of biomechanics, 45(16), 2931-2934 (2012-10-13)
Interest in soft tissue RadioStereometric Analysis (RSA) is rising. Previous studies have attempted to use this technique to analyse anterior cruciate ligament (ACL) graft constructs, and more recently, the movement of soft tissue as a precursor to RSA follow up
Yongyuan Guo et al.
International journal of molecular medicine, 31(3), 689-697 (2013-01-23)
Titanium (Ti) alloys of the β-type are highly attractive metallic materials for biomedical applications due to their low elastic modulus, high corrosion resistance and notable biocompatibility. A new β-type Ti35Nb2Ta3Zr alloy with a low Young's modulus of approximately 48 GPa was previously
In response.
Mariano Fernandez Fairen et al.
Spine, 38(4), 370-371 (2012-12-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service