Skip to Content
Merck
All Photos(2)

Key Documents

261599

Sigma-Aldrich

4-(Trimethylsilyl)morpholine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€37.90

€37.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€37.90

About This Item

Empirical Formula (Hill Notation):
C7H17NOSi
CAS Number:
Molecular Weight:
159.30
Beilstein:
1071480
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€37.90


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

160 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

C[Si](C)(C)N1CCOCC1

InChI

1S/C7H17NOSi/c1-10(2,3)8-4-6-9-7-5-8/h4-7H2,1-3H3

InChI key

JJOWIQMPCCUIGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for deoxynucleoside synthesis.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service