Skip to Content
Merck
All Photos(1)

Key Documents

259608

Sigma-Aldrich

m-Anisic hydrazide

98%

Synonym(s):

3-Methoxybenzoic hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CONHNH2
CAS Number:
Molecular Weight:
166.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 259608, please contact your local Merck office or dealer. Contact Technical Service

Quality Level

Assay

98%

form

solid

mp

93-95 °C (lit.)

SMILES string

COc1cccc(c1)C(=O)NN

InChI

1S/C8H10N2O2/c1-12-7-4-2-3-6(5-7)8(11)10-9/h2-5H,9H2,1H3,(H,10,11)

InChI key

VMZSDAQEWPNOIB-UHFFFAOYSA-N

Application

m-Anisic hydrazide has been used in the preparation of Schiff base ligands:
  • (5-chlorosalicylaldehyde furan-1-carbohydrazonato) hexamethylphosphoramide dioxomolybdenum (VI) dimethylformamide solvate[1]
  • 5-chlorosalicylaldehyde 3-methoxy-benzo hydrazone[1]
  • (5-chlorosalicylaldehyde 3-methoxy-benzylhydrazonato) dimethylsulfoxide dioxomolybdenum (VI)[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure studies and electrochemistry of molybdenum (VI) Schiff base complexes in the presence of different donor solvent molecules.
Ngan NK, et al.
Polyhedron, 30(17), 2922-2932 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service