Skip to Content
Merck
All Photos(1)

Key Documents

259047

Sigma-Aldrich

4-Indolyl acetate

99%

Synonym(s):

4-Acetoxyindole

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€73.90

€73.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€73.90

About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€73.90


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99%

form

solid

mp

100-102 °C (lit.)

functional group

ester

SMILES string

CC(=O)Oc1cccc2[nH]ccc12

InChI

1S/C10H9NO2/c1-7(12)13-10-4-2-3-9-8(10)5-6-11-9/h2-6,11H,1H3

InChI key

ZXDMUHFTJWEDEF-UHFFFAOYSA-N

Application

4-Indolyl acetate has been employed as standard in a protocol for the optimization of gradient reversed phase chromatographic peak capacity for separation of small molecules[1].
Reactant for preparation of:
  • Inhibitors of HIV-1 attachment
  • Prostanoid EP3 receptor antagonist, as a novel antiplatelet agent
  • Indolyl-nitroalkanes via N-bromosuccinimide catalyzed synthesis
  • Psilocybin as a medicinal agent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arianne Soliven et al.
Journal of chromatography. A, 1273, 57-65 (2012-12-25)
A general protocol for optimizing peak capacity for the separation of low molecular weight molecules under gradient elution conditions has not yet been developed. By studying the effects of gradient time, flow rate, temperature, final eluent composition, and column length

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service