Skip to Content
Merck
All Photos(1)

Key Documents

256803

Sigma-Aldrich

Diisobutylaluminum chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
[(CH3)2CHCH2]2AlCl
CAS Number:
Molecular Weight:
176.66
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4506 (lit.)

bp

152 °C/10 mmHg (lit.)

mp

−40 °C (lit.)

density

0.905 g/mL at 25 °C (lit.)

SMILES string

CC(C)C[Al](Cl)CC(C)C

InChI

1S/2C4H9.Al.ClH/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;1H/q;;+1;/p-1

InChI key

HQMRIBYCTLBDAK-UHFFFAOYSA-M

Application

Transmetalating reagent

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service