Skip to Content
Merck
All Photos(1)

Documents

253804

Sigma-Aldrich

2-Ethylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4CO2H
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

62-66 °C (lit.)

SMILES string

CCc1ccccc1C(O)=O

InChI

1S/C9H10O2/c1-2-7-5-3-4-6-8(7)9(10)11/h3-6H,2H2,1H3,(H,10,11)

InChI key

CGMMPMYKMDITEA-UHFFFAOYSA-N

Application

2-Ethylbenzoic acid has been employed:
  • as substrate for microbial asymmetric synthesis of (S)-3-methylphthalide
  • in synthesis of γ-lactones and bis(2-ethylbenzoyl)peroxide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microbial asymmetric syntheses of 3-alkylphthalide derivatives.
Kitayama T.
Tetrahedron Asymmetry, 8(22), 3765-3774 (1997)
Intramolecular hydrogen-atom transfer in 2-alkylbenzoyloxyl radicals as studied by transient absorption kinetics and product analyses on the photodecomposition of bis (2-alkylbenzoyl) peroxides.
Wang J, et al.
Bulletin of the Chemical Society of Japan, 68(4), 1213-1219 (1995)
Cynthia D Fast et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(11), 3085-3101 (2017-02-12)
Fear- and stress-induced activity in the amygdala has been hypothesized to influence sensory brain regions through the influence of the amygdala on neuromodulatory centers. To directly examine this relationship, we used optical imaging to observe odor-evoked activity in populations of
An improved method for the synthesis of ?-lactones using sodium bromate and sodium hydrogen sulfite.
Hayat S, et al.
Tetrahedron Letters, 42(9), 1647-1649 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service