Skip to Content
Merck
All Photos(1)

Key Documents

253707

Sigma-Aldrich

2-Methoxyphenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NCS
CAS Number:
Molecular Weight:
165.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
To inquire about this product 253707, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

liquid

refractive index

n20/D 1.6443 (lit.)

bp

264-266 °C (lit.)

density

1.196 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1N=C=S

InChI

1S/C8H7NOS/c1-10-8-5-3-2-4-7(8)9-6-11/h2-5H,1H3

InChI key

QKAOOWJWWKWWOZ-UHFFFAOYSA-N

General description

IR and Raman spectral measurements of liquid samples of 2-methoxyphenyl isothiocyanate have been investigated[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jayashree Yenagi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 91, 261-268 (2012-03-03)
IR and Raman spectral measurements in the region 3500-400/50 cm(-1) have been made for the liquid samples of 2-Methoxyphenyl isocyanate and 2-Methoxyphenyl isothiocyanate. A complete assignment of the measured bands has been proposed as aided by conformational and vibration analyses

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service