Skip to Content
Merck
All Photos(1)

Key Documents

24882

Sigma-Aldrich

5-Chloro-6-hydroxynicotinic acid

≥97.0%

Synonym(s):

5-Chloro-6-hydroxypyridine-3-carboxylicacid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO3
CAS Number:
Molecular Weight:
173.55
Beilstein:
127900
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0%

mp

≥300 °C (lit.)

SMILES string

OC(=O)c1cnc(O)c(Cl)c1

Looking for similar products? Visit Product Comparison Guide

Application

5-Chloro-6-hydroxynicotinic acid has been used in the preparation of triorganotin compounds via reaction with (R3Sn)2O (R=Ph, 2-ClC6H4CH2, 2-FC6H4CH2, 4-CNC6H4CH2).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Structure Characterization of some Triorganotin Esters of Heteroaromatic Carboxlic Acid and Crystal Structure of Triphenyltin Esters 5-Chloro-6-Hydroxynicotinic Acid.
Gao HT.
Advanced Materials Research, 466, 463-468 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service