Skip to Content
Merck
All Photos(1)

Key Documents

248398

Sigma-Aldrich

4,4′-Oxydianiline

purified by sublimation, 99%

Synonym(s):

4,4′-Diaminodiphenyl ether, 4-Aminophenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(C6H4NH2)2
CAS Number:
Molecular Weight:
200.24
Beilstein:
475735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

10 mmHg ( 240 °C)

Assay

99%

form

solid

purified by

sublimation

mp

188-192 °C (lit.)

SMILES string

Nc1ccc(Oc2ccc(N)cc2)cc1

InChI

1S/C12H12N2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2

InChI key

HLBLWEWZXPIGSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Oxydianiline is a flexible diamine and has been used to tailor the properties of octa(aminophenyl)-silsesquioxane cross-linked polyimide aerogels. It was also used in the preparation of novel polyimide hybrid nanocomposite containing polyhedral oligomeric silsesquioxane.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

426.2 °F - closed cup

Flash Point(C)

219 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuwang Duo et al.
Journal of nanoscience and nanotechnology, 13(2), 1356-1359 (2013-05-08)
A novel polyimide (PI) hybrid nanocomposite containing polyhedral oligomeric silsesquioxane (POSS) had been prepared by copolymerization of trisilanolphenyl-POSS, 4,4'-oxydianiline (ODA), and pyromellitic dianhydride (PMDA). The AO resistance of these PI/POSS hybrid films was tested in the ground-based AO simulation facility.
Haiquan Guo et al.
ACS applied materials & interfaces, 4(10), 5422-5429 (2012-09-08)
Combinations of rigid and flexible aromatic diamines were used to tailor the properties of octa(aminophenyl)-silsesquioxane (OAPS) cross-linked polyimide aerogels. 2,2'-Dimethylbenzidine (DMBZ) or p-phenylenediamine (PPDA) was used in combination with the more-flexible diamine, 4,4'-oxydianiline (ODA). The amount of rigid diamine was
Anders Lennartson et al.
Chirality, 27(7), 425-429 (2015-06-03)
We studied the spontaneous formation of chiral crystals of four diaryl ethers, 3-phenoxybenzaldehyde, 1; 1,3-dimethyl-2-phenoxybenzene, 2; di(4-aminophenyl) ether, 3; and di(p-tolyl) ether, 4. Compounds 1, 3, and 4 form conformationally chiral molecules in the solid state, while the chirality of
[Functional and morphologic changes in the myocardium caused by chemical damage].
M S Gnatiuk et al.
Gigiena i sanitariia, (2)(2), 54-57 (1990-02-01)
[Cardiotoxic action of 4,4-diaminodiphenyl ether].
V A Kondratiuk et al.
Gigiena i sanitariia, (6)(6), 31-34 (1986-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service