Skip to Content
Merck
All Photos(1)

Key Documents

244384

Sigma-Aldrich

1-Decyne

98%

Synonym(s):

Octylacetylene, n-Octylacetylene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€46.00
25 G
€106.00

€46.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€46.00
25 G
€106.00

About This Item

Linear Formula:
CH3(CH2)7C≡CH
CAS Number:
Molecular Weight:
138.25
Beilstein:
1236372
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010414
PubChem Substance ID:
NACRES:
NA.22

€46.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.427 (lit.)

bp

174 °C (lit.)

mp

−44 °C (lit.)

density

0.766 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC#C

InChI

1S/C10H18/c1-3-5-7-9-10-8-6-4-2/h1H,4-10H2,2H3

InChI key

ILLHQJIJCRNRCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds[1].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce D Phebus et al.
Physical chemistry chemical physics : PCCP, 15(40), 17647-17653 (2013-09-18)
Ruthenium nanoparticles were stabilized by the self-assembly of 1-decyne forming ruthenium-vinylidene interfacial bonds and further functionalized by metathesis reactions with 4-ethynyl-N,N-diphenylaniline (EDPA) and 9-vinylanthracene (VAN). Photoluminescence studies of the resulting bifunctionalized Ru(EDPA/VAN) nanoparticles showed that as both ligands were bound

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service