242217
Methyl Violet B base
Dye content 85 %
Synonym(s):
Solvent Violet 8
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Colour Index Number:
42535:1
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Recommended Products
composition
Dye content, 85%
λmax
580 nm
SMILES string
C\N=C1/C=CC(\C=C1)=C(\c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C
InChI
1S/C24H27N3/c1-25-21-12-6-18(7-13-21)24(19-8-14-22(15-9-19)26(2)3)20-10-16-23(17-11-20)27(4)5/h6-17H,1-5H3
InChI key
AMPCGOAFZFKBGH-UHFFFAOYSA-N
General description
Methyl Violet B base, also known as solvent violet 8, is a free base form of methyl violet. It belongs to the class of triarylmethane dyes. Methyl Violet B base is bright bluish violet in color and is widely used as a flexographic ink for paper bags.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Robert Leach
The Printing Ink Manual (2012)
R. L. Allen
Colour Chemistry (2013)
Niir Board
The Complete Technology Book On Dyes & Dye Intermediates (2003)
Oscar Díaz-Santana et al.
Journal of chromatography. A, 1515, 187-195 (2017-08-16)
An extraction and determination method is shown for the analysis of dyes and solvents present in two types of ballpoint pen inks that are deposited onto paper. Ink extracts are analysed using a combination of gas chromatography with mass spectrometry
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service